Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083907.2 | NP_001077376.2 | 3 | 540 | ||
NM_001127507.2 | NP_001120979.2 | 2 | 372 | |||
NM_017935.4 | NP_060405.4 | 3 | 771 | |||
XM_017008337.1 | XP_016863826.1 | 3 | 518 | |||
GenBank mRNA | AB063170.1 | - | 2 | 407 | ||
AK000713.1 | - | 3 | 487 | |||
AK091523.1 | - | 7 | 872 | |||
AK292133.1 | - | 3 | 615 | |||
AL834291.1 | - | 3 | 590 | |||
BC032241.1 | - | 3 | 639 | 5214147 | ||
DA946668.1 | - | 3 | 541 | |||
EU051376.1 | - | 2 | 110 |