Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACOXL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011511416.2 | XP_011509718.1 | 19 | 2499 | ||
XM_011511419.2 | XP_011509721.1 | 18 | 2409 | |||
GenBank mRNA | AK001904.1 | - | 13 | 1758 | ||
BC131514.1 | - | 12 | 1665 |