Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UEVLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040697.2 | NP_001035787.1 | 9 | 1118 | ||
NM_001261382.1 | NP_001248311.1 | 8 | 1052 | |||
NM_001261383.1 | NP_001248312.1 | 8 | 1052 | |||
NM_001261384.1 | NP_001248313.1 | 7 | 869 | |||
NM_001261385.1 | NP_001248314.1 | 9 | 1075 | |||
NM_001261386.1 | NP_001248315.1 | 6 | 647 | |||
NM_018314.4 | NP_060784.3 | 9 | 1118 | |||
XM_006718255.2 | XP_006718318.1 | 8 | 947 | |||
XM_011520201.2 | XP_011518503.1 | 5 | 2949 | |||
XM_017017985.1 | XP_016873474.1 | 8 | 1045 | |||
XM_017017986.1 | XP_016873475.1 | 8 | 925 | |||
GenBank mRNA | AF503350.1 | - | 9 | 1078 | ||
AK001930.1 | - | 9 | 1075 | |||
AK295501.1 | - | 7 | 797 | |||
AK296835.1 | - | 8 | 1012 | |||
AK301537.1 | - | 8 | 994 | |||
AK314521.1 | - | 9 | 1118 | |||
AK316191.1 | - | 8 | 925 | |||
BC064566.1 | - | 8 | 992 | 5576632 | ||
CD109744.1 | - | 6 | 646 | |||
CF138618.1 | - | 1 | 125 |