Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC29A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174098.1 | NP_001167569.1 | 6 | 1583 | ||
NM_018344.5 | NP_060814.4 | 6 | 1577 | |||
NR_033413.1 | 5 | 1494 | ||||
NR_033414.1 | 4 | 1267 | ||||
XM_006717910.3 | XP_006717973.1 | 6 | 1969 | |||
XM_017016377.1 | XP_016871866.1 | 5 | 1791 | |||
XM_017016378.1 | XP_016871867.1 | 4 | 1567 | |||
GenBank mRNA | AK002022.1 | - | 5 | 1328 | ||
AK304503.1 | - | 5 | 1465 | |||
AK316152.1 | - | 5 | 1479 | |||
AX775915.1 | - | 6 | 1577 | |||
AX775917.1 | - | 6 | 1577 | |||
AX775919.1 | - | 6 | 1577 | |||
AX775921.1 | - | 6 | 1577 | |||
AY358686.1 | - | 6 | 1555 | |||
BC000223.1 | - | 3 | 952 | |||
BC041575.1 | - | 5 | 1465 |