Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018951.3 | NP_061824.3 | 2 | 2326 | ||
NR_037939.1 | 2 | 1954 | ||||
GenBank mRNA | BC013971.2 | - | 2 | 2295 | 4107013 |