Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001290.1 | NP_001001290.1 | 5 | 564 | ||
NM_020041.2 | NP_064425.2 | 4 | 553 | |||
XM_006713968.3 | XP_006714031.1 | 4 | 665 | |||
XM_006713969.2 | XP_006714032.1 | 5 | 526 | |||
XM_011513856.2 | XP_011512158.1 | 4 | 690 | |||
XM_011513857.1 | XP_011512159.1 | 5 | 526 | |||
XM_011513858.1 | XP_011512160.1 | 5 | 728 | |||
XM_011513859.2 | XP_011512161.1 | 4 | 660 | |||
XM_011513860.2 | XP_011512162.1 | 4 | 660 | |||
XM_011513861.2 | XP_011512163.1 | 4 | 662 | |||
XM_011513862.2 | XP_011512164.1 | 3 | 537 | |||
XM_011513863.2 | XP_011512165.1 | 3 | 2216 | |||
XM_011513864.2 | XP_011512166.1 | 2 | 105 | |||
XM_011513865.2 | XP_011512167.1 | 4 | 664 | |||
XM_011513866.2 | XP_011512168.1 | 4 | 662 | |||
XM_011513868.2 | XP_011512170.1 | 4 | 681 | |||
XM_017008457.1 | XP_016863946.1 | 4 | 678 | |||
XM_017008458.1 | XP_016863947.1 | 4 | 663 | |||
XM_017008459.1 | XP_016863948.1 | 2 | 197 | |||
XM_017008460.1 | XP_016863949.1 | 3 | 2210 | |||
GenBank mRNA | AF210317.1 | - | 4 | 546 | ||
AF421859.1 | - | 2 | 241 | |||
AK310895.1 | - | 5 | 526 | |||
BC018897.2 | - | 5 | 564 | 3949549 | ||
BC110414.1 | - | 4 | 583 | 5762360 | ||
BI821650.1 | - | 4 | 559 | |||
CD629819.1 | - | 4 | 558 |