Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002027.2 | NP_002018.1 | 4 | 506 | ||
NR_033698.1 | 3 | 420 | ||||
GenBank mRNA | AK123002.1 | - | 4 | 893 | ||
AK292121.1 | - | 4 | 482 | |||
AL698961.1 | - | 2 | 189 | |||
BC017029.2 | - | 4 | 449 | |||
BC084566.1 | - | 4 | 466 | 6423445 | ||
BI913631.1 | - | 4 | 653 | |||
BM686906.1 | - | 3 | 399 | |||
BT009854.1 | - | 4 | 458 | |||
L00634.1 | - | 4 | 474 | |||
L10413.1 | - | 4 | 464 |