Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020346.2 | NP_065079.1 | 8, 9 | 1462 | ||
GenBank mRNA | AB032435.1 | - | 8, 9 | 1462 | ||
AK294502.1 | - | 7, 8 | 832 | |||
AK309131.1 | - | 6, 7 | 715 | |||
BC069629.1 | - | 8, 9 | 1208 | 7262471 | ||
BC069640.1 | - | 8, 9 | 1209 | 7262388 | ||
BC069646.1 | - | 8, 9 | 1209 | 7262400 |