Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V0A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020632.2 | NP_065683.2 | 10 | 1088 | ||
NM_130840.2 | NP_570855.2 | 9 | 985 | |||
NM_130841.2 | NP_570856.2 | 9 | 1043 | |||
XM_005250393.1 | XP_005250450.1 | 11 | 1435 | |||
XM_005250394.3 | XP_005250451.1 | 9 | 1347 | |||
GenBank mRNA | AF245517.1 | - | 10 | 1073 | ||
AK055789.1 | - | 5 | 476 | |||
AK292945.1 | - | 10 | 1088 | |||
BC109304.1 | - | 9 | 893 | 40035236 | ||
BC109305.1 | - | 9 | 893 | 40035237 |