Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308440.1 | NP_001295369.1 | 6, 7 | 881 | ||
NM_020828.1 | NP_065879.1 | 6, 7 | 881 | |||
XM_011526462.2 | XP_011524764.1 | 6, 7 | 877 | |||
XM_011526463.2 | XP_011524765.2 | 6, 7 | 1610 | |||
GenBank mRNA | AB037852.1 | - | 6, 7 | 881 | ||
AF507045.1 | - | 6, 7 | 881 | |||
AK289583.1 | - | 6, 7 | 866 | |||
BC028416.2 | - | 6, 7 | 876 | |||
BC099903.1 | - | 6, 7 | 876 | |||
BC127002.1 | - | 6, 7 | 866 | 40108311 | ||
BC127003.1 | - | 6, 7 | 866 | 40108312 |