Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002053.2 | NP_002044.2 | 7 | 1337 | ||
GenBank mRNA | AB208912.1 | - | 6 | 1902 | ||
AK291783.1 | - | 7 | 1185 | |||
AK298032.1 | - | 4 | 511 | |||
AK303893.1 | - | 7 | 1140 | |||
AY920436.1 | - | 6 | 1121 | |||
BC002666.2 | - | 7 | 1337 | 3606865 | ||
BT006847.1 | - | 6 | 1117 | |||
M55542.1 | - | 7 | 1185 |