Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC15A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145998.1 | NP_001139470.1 | 9 | 959 | ||
NM_021082.3 | NP_066568.3 | 10 | 1052 | |||
XM_005247722.3 | XP_005247779.1 | 10 | 936 | |||
XM_006713736.3 | XP_006713799.1 | 10 | 936 | |||
XM_017007074.1 | XP_016862563.1 | 10 | 936 | |||
GenBank mRNA | AK289820.1 | - | 10 | 936 | ||
AK302002.1 | - | 9 | 846 | |||
AK304189.1 | - | 9 | 844 | |||
BC038445.1 | - | 10 | 935 | |||
BC044572.1 | - | 10 | 936 | 5288920 | ||
S78203.1 | - | 10 | 929 |