Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021089.2 | NP_066575.2 | 4 | 2102 | ||
GenBank mRNA | AK093110.1 | - | 4 | 2102 | ||
AK225825.1 | - | 4 | 2098 | |||
BC039323.1 | - | 4 | 2060 | 5267031 | ||
M29581.1 | - | 3 | 1876 |