Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001118890.1 | NP_001112362.1 | 3 | 580 | ||
NM_001243658.1 | NP_001230587.1 | 3 | 998 | |||
NM_001243659.1 | NP_001230588.1 | 3 | 591 | |||
NM_002064.2 | NP_002055.1 | 3 | 1148 | |||
GenBank mRNA | AF069668.1 | - | 3 | 951 | ||
AK125636.1 | - | 2 | 3038 | |||
BC005304.1 | - | 3 | 433 | 3682161 | ||
BC010965.1 | - | 3 | 437 | 4333310 | ||
BC106075.1 | - | 3 | 436 | 4444172 | ||
BU949348.1 | - | 3 | 367 | |||
D21238.1 | - | 3 | 372 | |||
X76648.1 | - | 3 | 432 |