Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135146.1 | NP_001128618.1 | 7 | 1411 | ||
NM_001135147.1 | NP_001128619.1 | 7 | 1368 | |||
NM_001135148.1 | NP_001128620.1 | 6 | 848 | |||
NM_022154.5 | NP_071437.3 | 6 | 1533 | |||
XM_005263177.1 | XP_005263234.1 | 7 | 1320 | |||
XM_011532182.1 | XP_011530484.1 | 5 | 1346 | |||
XM_017008540.1 | XP_016864029.1 | 7 | 1328 | |||
XM_017008541.1 | XP_016864030.1 | 9 | 1705 | |||
XM_017008542.1 | XP_016864031.1 | 7 | 1407 | |||
GenBank mRNA | AB040120.1 | - | 6 | 1481 | ||
AF193052.1 | - | 7 | 1362 | |||
AK027652.1 | - | 6 | 848 | |||
AK304274.1 | - | 7 | 1320 | |||
BC012125.1 | - | 7 | 1387 | 4650026 |