Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MMS19 SIRNA Assays ›
Gene Symbol:
MMS19
Gene Name:
MMS19 homolog, cytosolic iron-sulfur assembly component
Gene Aliases:
MET18, MMS19L, hMMS19
Chromosome Location:
Chr.10: 97458324 - 97498794 on Build GRCh38
UniGene ID:
Hs.500721
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
109459
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MMS19
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289403.1
NP_001276332.1
26
2783
NM_001289404.1
NP_001276333.1
26
2848
NM_001289405.1
NP_001276334.1
28
2794
NM_022362.4
NP_071757.4
27
2912
XM_005270035.2
XP_005270092.1
26
2669
XM_005270041.1
XP_005270098.1
25
2557
XM_006717944.3
XP_006718007.2
27
2703
XM_006717945.2
XP_006718008.1
25
3245
XM_011540062.1
XP_011538364.1
26
2515
XM_011540063.2
XP_011538365.1
25
3243
XM_017016515.1
XP_016872004.1
28
2818
XM_017016516.1
XP_016872005.1
28
2815
XM_017016517.1
XP_016872006.1
27
2689
XM_017016518.1
XP_016872007.1
26
2574
XM_017016519.1
XP_016872008.1
26
2524
XM_017016520.1
XP_016872009.1
25
2412
XM_017016521.1
XP_016872010.1
25
2409
XM_017016522.1
XP_016872011.1
27
2810
XM_017016523.1
XP_016872012.1
26
2694
XM_017016524.1
XP_016872013.1
26
2666
XM_017016525.1
XP_016872014.1
25
2436
XM_017016526.1
XP_016872015.1
26
2672
XM_017016527.1
XP_016872016.1
26
3113
XM_017016528.1
XP_016872017.1
24
3084
XM_017016529.1
XP_016872018.1
26
3313
XM_017016530.1
XP_016872019.1
27
2832
XM_017016531.1
XP_016872020.1
25
2530
XM_017016532.1
XP_016872021.1
24
2307
XM_017016533.1
XP_016872022.1
24
2384
XM_017016534.1
XP_016872023.1
25
3240
XM_017016535.1
XP_016872024.1
24
3101
GenBank mRNA
AF007151.1
-
13
1374
AF319947.1
-
28
2760
AF357881.1
-
27
2711
AJ306408.1
-
27
2694
AK025496.1
-
14
1411
AK027710.1
-
24
2252
AK056244.1
-
16
1719
AK225312.1
-
27
2697
AK298995.1
-
27
2750
AK304287.1
-
26
2577
AK314374.1
-
27
2710
BC002692.1
-
16
1674
BC006575.2
-
19
2006
BC009396.2
-
10
978
BC080532.1
-
26
2627
4889876
BC117129.1
-
27
2710
40125680
BU942293.1
-
5
413
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
phosphorelay signal transduction system
DNA metabolic process
DNA repair
nucleotide-excision repair
transcription, DNA-templated
cellular response to DNA damage stimulus
chromosome segregation
response to hormone
iron-sulfur cluster assembly
positive regulation of transcription, DNA-templated
transcription coactivator activity
protein binding
enzyme binding
receptor signaling complex scaffold activity
estrogen receptor binding
protein binding, bridging
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-zp8kv:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0