Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LILRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003061.2 | 7 | 1910 | |||
GenBank mRNA | BC131616.1 | - | 7 | 1910 | ||
BG744172.1 | - | 3 | 394 |