Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025208.4 | NP_079484.1 | 7 | 1724 | ||
NM_033135.3 | NP_149126.1 | 7 | 1706 | |||
GenBank mRNA | AF113216.1 | - | 7 | 1458 | ||
AF335584.1 | - | 7 | 1533 | |||
AF336376.1 | - | 7 | 1527 | |||
AY027517.1 | - | 7 | 1444 | |||
AY027518.1 | - | 7 | 1426 | |||
BC030645.2 | - | 7 | 1786 | 4824526 |