Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320555.1 | NP_001307484.1 | 2 | 321 | ||
NM_001320556.1 | NP_001307485.1 | 2 | 271 | |||
NM_001320557.1 | NP_001307486.1 | 2 | 271 | |||
NM_001320558.1 | NP_001307487.1 | 2 | 271 | |||
NM_025231.2 | NP_079507.1 | 2 | 271 | |||
XM_017011324.1 | XP_016866813.1 | 2 | 1240 | |||
GenBank mRNA | AK025844.1 | - | 2 | 218 | ||
AK294017.1 | - | 2 | 218 | |||
AK309837.1 | - | 2 | 218 | |||
BC004255.1 | - | 2 | 192 | 3609511 | ||
CB158915.1 | - | 2 | 200 | |||
CX865974.1 | - | 1 | 161 | |||
DA093436.1 | - | 2 | 227 | |||
DB158261.1 | - | 2 | 271 |