Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317950.1 | NP_001304879.1 | 19 | 3915 | ||
NM_001317952.1 | NP_001304881.1 | 18 | 3473 | |||
NM_030767.5 | NP_110394.3 | 19 | 3860 | |||
XM_005252244.2 | XP_005252301.1 | 21 | 4032 | |||
XM_005252245.1 | XP_005252302.1 | 20 | 3890 | |||
XM_005252247.4 | XP_005252304.1 | 19 | 3911 | |||
XM_006717294.1 | XP_006717357.1 | 19 | 3844 | |||
XM_011519063.2 | XP_011517365.1 | 18 | 3523 | |||
XM_011519065.2 | XP_011517367.2 | 19 | 3503 | |||
XM_017015172.1 | XP_016870661.1 | 20 | 3630 | |||
GenBank mRNA | AF286341.1 | - | 9 | 1414 | ||
AK024431.1 | - | 17 | 2808 | |||
AK074040.1 | - | 18 | 2903 | |||
AK131082.2 | - | 19 | 3915 | |||
AK160382.1 | - | 19 | 3917 | |||
AL713755.1 | - | 2 | 2349 | |||
AL833220.1 | - | 16 | 2337 | |||
AY703039.1 | - | 20 | 3515 | |||
AY703040.1 | - | 18 | 3473 | |||
AY703043.1 | - | 18 | 3833 | |||
AY703044.1 | - | 17 | 3527 | |||
AY703045.1 | - | 10 | 1606 | |||
AY703046.1 | - | 6 | 732 | |||
BC042202.1 | - | 5 | 590 | |||
BC055285.1 | - | 19 | 3860 | 6143155 |