Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127496.1 | NP_001120968.1 | 2 | 3036 | ||
NM_001293289.1 | NP_001280218.1 | 3 | 3414 | |||
NM_001293290.1 | NP_001280219.1 | 2 | 3352 | |||
NM_030964.3 | NP_112226.2 | 3 | 3087 | |||
XM_011537685.2 | XP_011535987.1 | 3 | 3814 | |||
XM_017009910.1 | XP_016865399.1 | 2 | 4080 | |||
GenBank mRNA | AF227516.1 | - | 3 | 3027 | ||
AK226147.1 | - | 1 | 3459 |