Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032832.5 | NP_116221.3 | 7 | 1799 | ||
XM_011536194.2 | XP_011534496.1 | 6 | 1701 | |||
GenBank mRNA | AK027641.1 | - | 7 | 1481 | ||
AK299599.1 | - | 7 | 1042 | |||
BC043141.2 | - | 7 | 1800 | 5286304 |