Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033343.3 | NP_203129.1 | 6 | 1336 | ||
XM_011510105.2 | XP_011508407.1 | 6 | 1570 | |||
XM_011510106.2 | XP_011508408.1 | 6 | 1193 | |||
XM_011510108.2 | XP_011508410.1 | 6 | 976 | |||
XM_017002755.1 | XP_016858244.1 | 5 | 1688 | |||
GenBank mRNA | AB037683.1 | - | 3 | 654 | ||
AB055703.1 | - | 5 | 1039 | |||
AF179849.1 | - | 6 | 1340 | |||
AF282899.1 | - | 5 | 1035 | |||
AY053457.1 | - | 6 | 1128 | |||
BC011759.2 | - | 6 | 1147 | 3529089 | ||
KU178818.1 | - | 6 | 1104 | |||
KU178819.1 | - | 7 | 1155 |