Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040022.1 | NP_001035111.1 | 9 | 4081 | ||
NM_001040023.1 | NP_001035112.1 | 8 | 3989 | |||
NM_080792.2 | NP_542970.1 | 9 | 3748 | |||
XM_005260669.3 | XP_005260726.1 | 9 | 4143 | |||
XM_005260670.3 | XP_005260727.1 | 9 | 3759 | |||
XM_006723545.3 | XP_006723608.1 | 6 | 3330 | |||
XM_011529173.2 | XP_011527475.1 | 8 | 3748 | |||
GenBank mRNA | AB023430.1 | - | 9 | 3748 | ||
BC026692.1 | - | 9 | 3765 | 3916836 | ||
BC038510.2 | - | 8 | 3991 | 5261789 | ||
U06681.1 | - | 1 | 1839 |