Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322818.1 | NP_001309747.1 | 6 | 925 | ||
NM_080872.3 | NP_543148.2 | 6 | 1212 | |||
XM_011544400.2 | XP_011542702.1 | 6 | 1186 | |||
XM_011544401.2 | XP_011542703.1 | 6 | 1186 | |||
XM_017013044.1 | XP_016868533.1 | 6 | 1187 | |||
XM_017013045.1 | XP_016868534.1 | 6 | 1187 | |||
XM_017013046.1 | XP_016868535.1 | 6 | 1187 | |||
XM_017013048.1 | XP_016868537.1 | 6 | 2372 | |||
XM_017013049.1 | XP_016868538.1 | 6 | 2323 | |||
XM_017013050.1 | XP_016868539.1 | 7 | 1779 | |||
GenBank mRNA | AB055056.1 | - | 6 | 924 | ||
AK122615.1 | - | 2 | 279 | |||
AY358147.1 | - | 6 | 1212 |