Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EYA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282560.1 | NP_001269489.1 | 11 | 1070 | ||
NM_001282561.1 | NP_001269490.1 | 11 | 1070 | |||
NM_001282562.1 | NP_001269491.1 | 11 | 1128 | |||
NM_001990.3 | NP_001981.2 | 12 | 1208 | |||
NR_104214.1 | 12 | 1208 | ||||
XM_006710449.3 | XP_006710512.1 | 13 | 1244 | |||
XM_011540999.2 | XP_011539301.1 | 13 | 1247 | |||
XM_011541001.2 | XP_011539303.1 | 12 | 1205 | |||
XM_011541003.2 | XP_011539305.1 | 12 | 1107 | |||
XM_011541004.2 | XP_011539306.1 | 12 | 1104 | |||
XM_017000652.1 | XP_016856141.1 | 13 | 1248 | |||
XM_017000653.1 | XP_016856142.1 | 14 | 1368 | |||
XM_017000654.1 | XP_016856143.1 | 12 | 1205 | |||
XM_017000655.1 | XP_016856144.1 | 13 | 1361 | |||
XM_017000656.1 | XP_016856145.1 | 12 | 1108 | |||
XM_017000657.1 | XP_016856146.1 | 11 | 1100 | |||
XM_017000658.1 | XP_016856147.1 | 11 | 1125 | |||
XM_017000659.1 | XP_016856148.1 | 10 | 984 | |||
GenBank mRNA | AK289805.1 | - | 12 | 1167 | ||
AK295745.1 | - | 11 | 1064 | |||
AK298129.1 | - | 11 | 1125 | |||
AK303664.1 | - | 10 | 984 | |||
BC029500.1 | - | 12 | 1168 | 5272319 | ||
BC041667.1 | - | 11 | 997 | 5743906 | ||
BX648945.1 | - | 12 | 1177 | |||
U81602.1 | - | 7 | 807 |