Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286808.1 | NP_001273737.1 | 10 | 2430 | ||
NM_130809.4 | NP_570721.1 | 9 | 4348 | |||
XM_006714529.3 | XP_006714592.1 | 9 | 4556 | |||
GenBank mRNA | AJ441110.3 | - | 10 | 2381 | ||
AJ515158.1 | - | 10 | 2364 | |||
AJ515428.1 | - | 9 | 4299 | |||
AJ515429.1 | - | 9 | 4299 | |||
AK090486.1 | - | 8 | 4173 | |||
AL832056.1 | - | 9 | 4312 |