Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138800.1 | NP_620155.1 | 7 | 1457 | ||
GenBank mRNA | BC015353.1 | - | 7 | 1457 | 3445177 |
Gene Symbol: TRIM43B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164464.1 | NP_001157936.1 | 7 | 1451 | ||
XM_011511669.1 | XP_011509971.1 | 7 | 1395 |