Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144660.2 | NP_653261.1 | 5 | 1174 | ||
GenBank mRNA | AK057811.1 | - | 5 | 1102 | ||
EU668332.1 | - | 5 | 1102 | |||
GQ472208.1 | - | 5 | 1102 |