Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144976.3 | NP_659413.1 | 4 | 2200 | ||
GenBank mRNA | AK092760.1 | - | 4 | 2161 | ||
AL834146.1 | - | 4 | 5460 | |||
BC028367.1 | - | 4 | 2137 | |||
BC065023.1 | - | 4 | 2110 | 6044821 | ||
BC136728.1 | - | 4 | 2036 | 9020718 | ||
BC136729.1 | - | 4 | 2036 | 9020719 | ||
BX647959.1 | - | 4 | 2172 |