Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297743.1 | NP_001284672.1 | 2 | 783 | ||
NM_145007.3 | NP_659444.2 | 5 | 1633 | |||
GenBank mRNA | AK310394.1 | - | 5 | 1310 | ||
AY095145.1 | - | 3 | 1021 | |||
AY154466.1 | - | 2 | 922 | |||
AY168967.1 | - | 3 | 1010 | |||
AY168968.1 | - | 2 | 682 | |||
AY226382.1 | - | 2 | 922 | |||
BC034730.1 | - | 3 | 1010 | 4341197 |