Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145214.2 | NP_660215.1 | 6 | 2427 | ||
XM_011544285.2 | XP_011542587.1 | 5 | 2263 | |||
XM_017002412.1 | XP_016857901.1 | 6 | 2491 | |||
GenBank mRNA | AF220125.1 | - | 1 | 992 | ||
AF327056.1 | - | 6 | 2426 | |||
AK097825.1 | - | 6 | 1956 | |||
AK226119.1 | - | 5 | 5507 | |||
AL360165.1 | - | 1 | 958 | |||
BC011629.2 | - | 4 | 1519 | |||
BC069227.1 | - | 6 | 2418 | 4875080 |