Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002032.2 | NP_002023.2 | 4 | 669 | ||
GenBank mRNA | AB062402.1 | - | 4 | 622 | ||
AF088851.1 | - | 4 | 476 | |||
AK311818.1 | - | 4 | 640 | |||
AY258285.1 | - | 1227 | ||||
BC000857.1 | - | 4 | 628 | 3459353 | ||
BC001399.2 | - | 4 | 597 | 3051458 | ||
BC011359.2 | - | 4 | 619 | 4157296 | ||
BC013724.2 | - | 4 | 622 | 3857790 | ||
BC015156.1 | - | 4 | 492 | 3883694 | ||
BC016009.1 | - | 4 | 644 | 4666545 | ||
BC016857.1 | - | 4 | 588 | 3848510 | ||
BC020300.1 | - | 4 | 605 | |||
BC035441.1 | - | 4 | 628 | |||
BC063514.1 | - | 4 | 611 | 3882345 | ||
BC066341.1 | - | 5 | 546 | 5288688 | ||
BC066961.1 | - | 4 | 642 | 4796312 | ||
BC070494.1 | - | 5 | 583 | |||
BC073750.1 | - | 4 | 626 | 6256153 | ||
BC104643.1 | - | 4 | 644 | 4043391 | ||
BC105802.1 | - | 5 | 621 | |||
L20941.1 | - | 4 | 642 | |||
M11146.1 | - | 4 | 511 | |||
M12937.1 | - | 4 | 460 | |||
M15383.1 | - | 1 | 53 | |||
M97164.1 | - | 4 | 563 | |||
X00318.1 | - | 4 | 525 |
Gene Symbol: FTH1P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | J04755.1 | - | 1 | 1556 |