Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLHC1 SIRNA Assays ›
Gene Symbol:
CLHC1
Gene Name:
clathrin heavy chain linker domain containing 1
Gene Aliases:
C2orf63
Chromosome Location:
Chr.2: 55172547 - 55232563 on Build GRCh38
UniGene ID:
Hs.347014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
110153
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLHC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135598.1
NP_001129070.1
7, 8
908
NM_152385.2
NP_689598.2
9, 10
1359
XM_005264136.2
XP_005264193.1
8, 9
1227
XM_006711937.3
XP_006712000.2
6, 7
666
XM_006711938.3
XP_006712001.1
3, 4
286
XM_006711939.3
XP_006712002.1
2, 3
158
XM_011532519.2
XP_011530821.1
9, 10
4252
XM_011532520.2
XP_011530822.1
9, 10
1530
XM_011532521.2
XP_011530823.1
10, 11
1686
XM_011532522.2
XP_011530824.1
10, 11
1673
XM_011532523.2
XP_011530825.1
9, 10
1762
XM_011532524.1
XP_011530826.1
8, 9
1144
XM_011532525.2
XP_011530827.1
8, 9
1210
XM_011532527.2
XP_011530829.1
9, 10
4854
XM_011532528.1
XP_011530830.1
8, 9
1754
XM_011532529.2
XP_011530831.1
7, 8
1713
XM_011532530.1
XP_011530832.1
7, 8
1474
XM_011532534.1
XP_011530836.1
7, 8
1093
XM_011532535.2
XP_011530837.1
5, 6
603
XM_017003333.1
XP_016858822.1
7, 8
4054
XM_017003334.1
XP_016858823.1
7, 8
1332
XM_017003335.1
XP_016858824.1
7, 8
4422
XM_017003336.1
XP_016858825.1
8, 9
1843
XM_017003337.1
XP_016858826.1
6, 7
932
XM_017003338.1
XP_016858827.1
6, 7
910
XM_017003340.1
XP_016858829.1
7, 8
8354
XM_017003341.1
XP_016858830.1
7, 8
1839
XM_017003342.1
XP_016858831.1
6, 7
1340
XM_017003343.1
XP_016858832.1
6, 7
1556
XM_017003344.1
XP_016858833.1
6, 7
1644
XM_017003345.1
XP_016858834.1
5, 6
1514
XM_017003347.1
XP_016858836.1
4, 5
1382
XM_017003349.1
XP_016858838.1
4, 5
3801
XM_017003350.1
XP_016858839.1
4, 5
1312
GenBank mRNA
AK315683.1
-
9, 10
1317
BC029502.1
-
9, 10
1359
5270720
BC036908.1
-
7, 8
908
5166064
Back To Top
More Information
Panther Classification:
Molecular Function -
vesicle coat protein
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0