Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258025.1 | NP_001244954.1 | 4 | 788 | ||
NM_152620.2 | NP_689833.1 | 3 | 614 | |||
XM_011531683.2 | XP_011529985.1 | 2 | 608 | |||
GenBank mRNA | AK093201.1 | - | 3 | 615 | ||
BC100983.1 | - | 2 | 504 | 40005592 | ||
BC100984.1 | - | 2 | 504 | 40005593 | ||
BC100985.1 | - | 2 | 504 | 40005594 | ||
BC100986.2 | - | 3 | 678 | 40005597 |