Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIBADH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152740.3 | NP_689953.1 | 7, 8 | 1073 | ||
XM_017011719.1 | XP_016867208.1 | 8, 9 | 1103 | |||
GenBank mRNA | AB050000.1 | - | 7, 8 | 947 | ||
AF529362.1 | - | 7, 8 | 860 | |||
AK316605.1 | - | 7, 8 | 878 | |||
BC013855.1 | - | 4, 5 | 446 | |||
BC020167.1 | - | 7, 8 | 934 | |||
BC032324.1 | - | 7, 8 | 958 | 3890576 |