Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317721.1 | NP_001304650.1 | 11, 12 | 1666 | ||
NM_152892.2 | NP_690852.1 | 11, 12 | 1648 | |||
XM_017011845.1 | XP_016867334.1 | 11, 12 | 1603 | |||
GenBank mRNA | AK290967.1 | - | 11, 12 | 1547 | ||
AK313771.1 | - | 11, 12 | 1531 | |||
AL133057.1 | - | 11, 12 | 1598 | |||
BC009436.1 | - | 10, 11 | 1339 | |||
BC018769.1 | - | 10, 11 | 1178 | |||
BC030547.1 | - | 11, 12 | 1588 | 5170588 |