Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XKR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173683.3 | NP_775954.2 | 3 | 1341 | ||
GenBank mRNA | AY534244.1 | - | 3 | 1341 | ||
BC024146.2 | - | 3 | 644 | 3678468 |