Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031679.2 | NP_001026849.1 | 3, 4 | 457 | ||
NM_001193460.1 | NP_001180389.1 | 4, 5 | 621 | |||
NM_001193461.1 | NP_001180390.1 | 3, 4 | 319 | |||
NM_198080.3 | NP_932346.1 | 2, 3 | 330 | |||
GenBank mRNA | AK293084.1 | - | 3, 4 | 427 | ||
AK299065.1 | - | 2, 3 | 269 | |||
AK316365.1 | - | 2, 3 | 301 | |||
AL833622.1 | - | 3, 4 | 375 | |||
AY358229.1 | - | 2, 3 | 265 | |||
BC040053.1 | - | 4, 5 | 522 | 6025598 | ||
BX500552.1 | - | 3, 4 | 321 | |||
BX648776.1 | - | 3, 4 | 376 | |||
DC357690.1 | - | 2, 3 | 330 |