Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QRFPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198179.2 | NP_937822.2 | 2 | 869 | ||
XM_017008693.1 | XP_016864182.1 | 2 | 872 | |||
GenBank mRNA | AB109629.1 | - | 2 | 490 | ||
AK309337.1 | - | 2 | 902 | |||
BC128132.1 | - | 2 | 490 | |||
BC128133.1 | - | 2 | 490 | 40112420 | ||
JF810892.1 | - | 2 | 490 |
Set Membership: |
G-protein coupled receptor |