Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000015.2 | NP_000006.2 | 2 | 609 | ||
XM_017012938.1 | XP_016868427.1 | 2 | 960 | |||
GenBank mRNA | BC015878.1 | - | 2 | 571 | 4716636 | |
BC067218.1 | - | 2 | 606 | 4722596 | ||
CR407631.1 | - | 1 | 502 | |||
D90040.1 | - | 2 | 543 | |||
D90042.1 | - | 2 | 609 |