Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGXT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000030.2 | NP_000021.1 | 7, 8 | 902 | ||
GenBank mRNA | AB292648.1 | - | 7, 8 | 803 | ||
AF348451.1 | - | 7, 8 | 693 | |||
AK292754.1 | - | 7, 8 | 823 | |||
AK315331.1 | - | 7, 8 | 850 | |||
BC132819.1 | - | 7, 8 | 899 | 40146841 | ||
D13368.1 | - | 7, 8 | 889 | |||
X53414.1 | - | 7, 8 | 903 | |||
X56092.1 | - | 7, 8 | 802 |