Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000163.4 | NP_000154.1 | 7 | 903 | ||
NM_001242399.2 | NP_001229328.1 | 7 | 869 | |||
NM_001242400.2 | NP_001229329.1 | 8 | 1154 | |||
NM_001242401.3 | NP_001229330.1 | 8 | 862 | |||
NM_001242402.2 | NP_001229331.1 | 7 | 831 | |||
NM_001242403.2 | NP_001229332.1 | 7 | 1039 | |||
NM_001242404.2 | NP_001229333.1 | 7 | 864 | |||
NM_001242405.2 | NP_001229334.1 | 7 | 759 | |||
NM_001242406.2 | NP_001229335.1 | 6 | 1125 | |||
NM_001242460.1 | NP_001229389.1 | 5 | 656 | |||
NM_001242462.1 | NP_001229391.1 | 6 | 722 | |||
GenBank mRNA | BC136496.1 | - | 5 | 604 | 9020485 | |
X06562.1 | - | 7 | 754 |