Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000481.3 | NP_000472.2 | 2, 3 | 496 | ||
NM_001164710.1 | NP_001158182.1 | 2, 3 | 496 | |||
NM_001164711.1 | NP_001158183.1 | 1, 2 | 328 | |||
NM_001164712.1 | NP_001158184.1 | 2, 3 | 496 | |||
NR_028435.1 | 2, 3 | 482 | ||||
GenBank mRNA | AK091738.1 | - | 2, 3 | 285 | ||
AK096062.1 | - | 2, 3 | 333 | |||
AK290600.1 | - | 2, 3 | 276 | |||
AK293481.1 | - | 1, 2 | 101 | |||
AK293594.1 | - | 1, 2 | 358 | |||
AK294592.1 | - | 2, 3 | 169 | |||
AK296177.1 | - | 2, 3 | 287 | |||
AK301072.1 | - | 2, 3 | 262 | |||
BC007546.1 | - | 2, 3 | 260 | |||
D13811.1 | - | 2, 3 | 397 | |||
DC387800.1 | - | 2, 3 | 496 |