Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002944.2 | NP_002935.2 | 43 | 7018 | ||
XM_006715548.3 | XP_006715611.1 | 44 | 7110 | |||
XM_011536049.2 | XP_011534351.1 | 44 | 7156 | |||
XM_011536050.2 | XP_011534352.1 | 44 | 7153 | |||
XM_011536051.2 | XP_011534353.1 | 43 | 7129 | |||
XM_011536052.2 | XP_011534354.1 | 44 | 7114 | |||
XM_011536053.2 | XP_011534355.1 | 44 | 6982 | |||
XM_017011172.1 | XP_016866661.1 | 43 | 7087 | |||
XM_017011173.1 | XP_016866662.1 | 43 | 7186 | |||
GenBank mRNA | BC144556.1 | - | 41 | 6200 | ||
M13880.1 | - | 10 | 1262 | |||
M34353.1 | - | 43 | 7025 |