Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000965.4 | NP_000956.2 | 4 | 1000 | ||
NM_001290216.2 | NP_001277145.1 | 7 | 1043 | |||
NM_001290217.1 | NP_001277146.1 | 4 | 458 | |||
NM_001290266.1 | NP_001277195.1 | 4 | 643 | |||
NM_001290276.1 | NP_001277205.1 | 4 | 1000 | |||
NM_001290277.1 | NP_001277206.1 | 4 | 1000 | |||
NM_001290300.1 | NP_001277229.1 | 4 | 464 | |||
NM_016152.3 | NP_057236.1 | 4 | 643 | |||
NR_110893.1 | 4 | 1000 | ||||
GenBank mRNA | AB307688.1 | - | 4 | 531 | ||
AF157483.1 | - | 4 | 642 | |||
AK292032.1 | - | 4 | 550 | |||
AK299756.1 | - | 4 | 618 | |||
AY501390.1 | - | 4 | 432 | |||
BC050415.2 | - | 2 | 226 | |||
BC060794.1 | - | 4 | 682 | 30341884 | ||
CD014020.1 | - | 4 | 547 | |||
CD014022.1 | - | 4 | 547 | |||
CR536558.1 | - | 2 | 195 | |||
DA882771.1 | - | 4 | 458 | |||
DQ083391.1 | - | 4 | 464 | |||
HQ692863.1 | - | 2 | 195 | |||
HQ692864.1 | - | 4 | 531 | |||
X07282.1 | - | 4 | 874 | |||
Y00291.1 | - | 4 | 852 |