Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004327.3 | NP_004318.3 | 6 | 2497 | ||
NM_021574.2 | NP_067585.2 | 6 | 2497 | |||
GenBank mRNA | AB209991.1 | - | 6 | 2246 | ||
AB742171.1 | - | 2 | 214 | |||
AK303241.1 | - | 6 | 931 | |||
AY533677.1 | - | 4 | 353 | |||
BC066122.1 | - | 6 | 2328 | |||
X02596.1 | - | 6 | 2389 | |||
Y00661.1 | - | 6 | 2497 |