Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192.2 | NP_001183.2 | 3 | 680 | ||
GenBank mRNA | AY684975.1 | - | 2 | 417 | ||
BC058291.1 | - | 3 | 603 | 6736094 | ||
EF152353.1 | - | 3 | 459 | |||
EF152354.1 | - | 3 | 369 | |||
EF152355.1 | - | 2 | 427 | |||
Z14954.1 | - | 3 | 528 | |||
Z29575.1 | - | 3 | 680 |