Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207019.2 | NP_001193948.2 | 5 | 386 | ||
NM_001220500.1 | NP_001207429.1 | 6 | 522 | |||
NM_002002.4 | NP_001993.2 | 6 | 518 | |||
XM_005272462.4 | XP_005272519.1 | 6 | 1123 | |||
GenBank mRNA | AK310071.1 | - | 6 | 507 | ||
AK310707.1 | - | 6 | 487 | |||
BC014108.2 | - | 6 | 483 | 4309266 | ||
BC062591.1 | - | 6 | 476 | 5216918 | ||
BC064417.1 | - | 5 | 800 | |||
M14766.1 | - | 6 | 490 | |||
M15059.1 | - | 6 | 518 | |||
X04772.1 | - | 6 | 518 |