Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002027.2 | NP_002018.1 | 2 | 264 | ||
GenBank mRNA | AK123002.1 | - | 2 | 651 | ||
AK292121.1 | - | 2 | 240 | |||
BC017029.2 | - | 2 | 207 | |||
BC084566.1 | - | 2 | 224 | 6423445 | ||
BI913631.1 | - | 2 | 410 | |||
BT009854.1 | - | 2 | 216 | |||
L00634.1 | - | 2 | 232 | |||
L10413.1 | - | 2 | 222 |